Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VIGNVQEKAM KLHNASNSEM VLY
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3' phosphoadenosine 5' phosphosulfate transporter 2; 3'-phosphoadenosine 5'-phosphosulfate transporter; Adenosine 3'-phospho 5'-phosphosulfate transporter 2; PAPS transporter 2; solute carrier family 35 (adenosine 3'-phospho 5'-phosphosulfate transporter), member B3; Solute carrier family 35 member B3; solute carrier family 35, member B3
Gene Aliases: C6orf196; CGI-19; PAPST2; SLC35B3
UniProt ID: (Human) Q9H1N7
Entrez Gene ID: (Human) 51000
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support