Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSVTKSTEGP QGAVAIKLDL MSPPESAKKL ENKDSTFLDE SPSESAGLKK TKGITVFQAL IHLVKGNMGT G
regulates pH-dependent, Na(+)-independent uptake of glycine, proline, and alpha(methyl)aminoisobutyric acid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16051; MGC119658; MGC119660; Proton-coupled amino acid transporter 2; Proton/amino acid transporter 2; solute carrier family 36 (proton/amino acid symporter), member 2; Solute carrier family 36 member 2; Tramdorin-1; Transmembrane domain rich protein 1
Gene Aliases: PAT2; SLC36A2; TRAMD1
UniProt ID: (Human) Q495M3
Entrez Gene ID: (Human) 153201
Molecular Function:
amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support