Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: RKTFSFVMPS LVEEIPLDKD DLGFITSSQS AAYAISKFVS GVLSDQMSA
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glucose-5-phosphate transporter; glucose-6-phosphatase, transport (glucose) protein 3; glucose-6-phosphatase, transport (glucose-6-phosphate) protein 1; glucose-6-phosphatase, transport (phosphate/pyrophosphate) protein 2; Glucose-6-phosphate exchanger SLC37A4; Glucose-6-phosphate translocase; MGC15729; microsomal glucose-6-phosphate transporter; solute carrier family 37 (glucose-6-phosphate transporter), member 4; Solute carrier family 37 member 4; Transformation-related gene 19 protein; TRG-19
Gene Aliases: G6PT; G6PT1; G6PT2; G6PT3; GSD1b; GSD1c; GSD1d; PRO0685; SLC37A4; TRG-19; TRG19
UniProt ID: (Human) O43826
Entrez Gene ID: (Human) 2542
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support