Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
1 µg/mL of MA5-45530 was sufficient for detection of SNAT1 in 20 µg of lysates from neocortical neurons cultured under amino acid starvation conditions and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 50kDa.
This antibody was formerly sold as clone S104-32.
Amino acid transporters play essential roles in the uptake of nutrients, production of energy, chemical metabolism, detoxification, and neurotransmitter cycling. SLC38A1 is an important transporter of glutamine, an intermediate in the detoxification of ammonia and the production of urea. Glutamine serves as a precursor for the synaptic transmitter, glutamate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amino acid transporter A1; amino acid transporter system A1; Glutamine transporter; MNat2; N-system amino acid transporter 2; rATA1; S38A1; SNAT 1; Sodium-coupled neutral amino acid symporter 1; sodium-coupled neutral amino acid transporter 1; Solute carrier family 38 member 1; solute carrier family 38, member 1; System A amino acid transporter 1; System A transporter 2; System N amino acid transporter 1
Gene Aliases: AA408026; AA409865; AL022800; ATA1; AU015942; GlnT; NAT2; Sa2; SAT1; SLC38A1; SNAT1
UniProt ID: (Human) Q9H2H9, (Rat) Q9JM15, (Mouse) Q8K2P7
Entrez Gene ID: (Human) 81539, (Rat) 170567, (Mouse) 105727
Molecular Function:
amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support