Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MGSNSGQAGR HIYKSLADDG PFDSVEPPKR PTSRLIMHSM AMFGREFCYA VEAAYVTPVL LSVGLPSSLY SIVWFLSPIL GFLLQPVVGS ASDHCRSRWG RRRPYILTLG VMMLVGMALY LNGATVVAAL IANPRRKLVW AISVTMIGVV LFDFAADFID GPIKAYLFDV CSHQDKEKGL HYHALFTDSQ GNDIKVTAES TGEHASSLPL PLHQPPHWMD GLPVQHAVLH RFHGPDCVPR GSL
SLC45A2 is a melanocyte differentiation antigen that is expressed in a high percentage of melanoma cell lines. A similar sequence gene in medaka, 'B, ' encodes a transporter that mediates melanin synthesis. Mutations in this gene are a cause of oculocutaneous albinism type 4. The protein encoded by this gene encodes a melanocyte differentiation antigen that is expressed in a high percentage of melanoma cell lines. A similar sequence gene in medaka, 'B, ' encodes a transporter that mediates melanin synthesis. Mutations in this gene are a cause of oculocutaneous albinism type 4. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Melanoma antigen AIM1; Membrane-associated transporter protein; Protein AIM-1; Solute carrier family 45 member 2; solute carrier family 45, member 2; underwhite
Gene Aliases: 1A1; AIM1; MATP; OCA4; SHEP5; SLC45A2
UniProt ID: (Human) Q9UMX9
Entrez Gene ID: (Human) 51151
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support