Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: SLC5A7.
Acetylcholine functions as a neurotransmitter in both the central and peripheral nervous systems of all vertebates, and is the principle neurotransmitter used at the neuromuscular junction. Presynaptic synthesis of acetylcholine requires a steady supply of choline, which is acquired by a plasma membrane high affinity choline transporter (CHT).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CHT; hCHT; hCHT1; Hemicholinium-3-sensitive choline transporter; High affinity choline transporter 1; high affinity choline transporter; hemicholinium-3-sensitive choline transporter; MGC126299; MGC126300; rCHT1; solute carrier family 5 (choline transporter), member 7; solute carrier family 5 (sodium/choline cotransporter), member 7; Solute carrier family 5 member 7; solute carrier family 5, member 7
Gene Aliases: CHT; CHT1; HMN7A; SLC5A7
UniProt ID: (Human) Q9GZV3, (Mouse) Q8BGY9, (Rat) Q9JMD7
Entrez Gene ID: (Human) 60482, (Mouse) 63993, (Rat) 85426
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support