Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
Keep as concentrated solution.
Predicted reactivity: Human (98%), Mouse (100%), Rat (100%), Xenopus laevis (85%), Rhesus Monkey (98%), Bovine (94%).
Positive Control: rat cerebellum.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GAT-1 (SLC6A1) The protein encoded by SLC6A1 is a gamma-aminobutyric acid (GABA) transporter that localizes to the plasma membrane. The encoded protein removes GABA from the synaptic cleft, restoring it to presynaptic terminals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA T-1; GABA T1; GABA transporter protein; GABAT1; gamma-aminobutyric acid (GABA-A) transporter 1; GAT-1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; Solute carrier family 6 member 1
Gene Aliases: A730043E01; GABATHG; GABATR; Gabt; Gabt1; GAT-1; Gat1; Slc6a1; XT-1; Xtrp1
UniProt ID: (Rat) P23978, (Mouse) P31648
Entrez Gene ID: (Rat) 79212, (Mouse) 232333
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support