Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total SLC6A11.
Slc6a11 encode GABA transporters that are sodium- and chloride-dependent members of the solute carrier family 6 (SLC6) and mediate the rapid removal of GABA and maintain low extracellular levels. Four 12-TM domain transporters have been identified and localized to neurons and glia. The protein encoded by the Slc6a11 gene is a sodium-dependent transporter that uptakes gamma-aminobutyric acid (GABA), an inhibitory neurotransmitter, which ends the GABA neurotransmission. Defects in the Slc6a11 gene may result in epilepsy, behavioral problems, or intellectual problems. Two transcript variants encoding different isoforms have been found for the Slc6a11 gene. Diseases associated with SLC6A11 include Tricuspid Valve Stenosis and Pulmonary Valve Insufficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA transporter 3; GABA transporter GAT-3; gamma-aminobutyric acid (GABA-A) transporter 4; GAT-3; GAT-4; Sodium- and chloride-dependent GABA transporter 3; Sodium- and chloride-dependent GABA transporter 4; solute carrier family 6 (neurotransmitter transporter), member 11; solute carrier family 6 (neurotransmitter transporter, GABA), member 11; Solute carrier family 6 member 11
Gene Aliases: D930045G19Rik; E130202I16Rik; GABT3; Gabt4; GAT-3; Gat-4; Gat-b; GAT3; GAT4; SLC6A11
UniProt ID: (Human) P48066, (Rat) P31647, (Mouse) P31650
Entrez Gene ID: (Human) 6538, (Rat) 79213, (Mouse) 243616
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support