Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Antibody detects endogenous levels of total SLC6A13.
Slc6a13 catalyzes sodium and chloride dependent transport of gamma-aminobutyric acid (GABA) to terminate GABA mediated synaptic activity. Slc6a13 is also a high-affinity gamma-aminobutyric acid (GABA) transporter, terminates biological actions of GABA by reuptake into cell, and its reuptake function utilizes sodium and chloride concentration gradients. Further, Slc6a13 may also aid in beta-alanine transport. Diseases associated with SLC6A13 include Traumatic Glaucoma and Neovascular Glaucoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA transport protein; GABA transporter 2; Gabt4; gamma-aminobutyric acid (GABA-A) transporter 3; GAT-2; GAT-3; Sodium- and chloride-dependent GABA transporter 2; Sodium- and chloride-dependent GABA transporter 3; solute carrier family 6 (neurotransmitter transporter), member 13; solute carrier family 6 (neurotransmitter transporter, GABA), member 13; Solute carrier family 6 member 13
Gene Aliases: Gabt2; Gabt3; GAT-2; Gat-3; GAT2; GAT3; SLC6A13
UniProt ID: (Human) Q9NSD5, (Mouse) P31649, (Rat) P31646
Entrez Gene ID: (Human) 6540, (Mouse) 14412, (Rat) 171163
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support