Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene is a member of the gamma-aminobutyric acid (GABA) neurotransmitter gene family and encodes a high-affinity mammalian brain L-proline transporter protein. This transporter protein differs from other sodium-dependent plasma membrane carriers by its pharmacological specificity, kinetic properties, and ionic requirements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain-specific L-proline transporter; L-proline transporter; proline transporter; slc6a7; Sodium-dependent proline transporter; solute carrier family 6 (neurotransmitter transporter, L-proline), member 7; Solute carrier family 6 member 7
Gene Aliases: AW455934; PROT; SLC6A7
UniProt ID: (Human) Q99884, (Rat) P28573, (Mouse) Q6PGE7
Entrez Gene ID: (Human) 6534, (Rat) 117100, (Mouse) 240332
Molecular Function: primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support