Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MEAARTAVLR VKRKRSAEPA EALVLACKRL RSDAVESAAQ KTSEDLERAA ENNVFHLVAT VCSQEEPVQP LLREVLRPSR DSQQRVRRNL RASAREVRQE GRYRVLSSRR SLGTTSSGQE SEYTPGNPEA AGNSGFQLLD LVHEEGEPEA ASAGSCKTSD PDVILCNSVE LIRERLTVSE DGPGVRRQEE QKHDDYVYDI YYLETATPGW IENILSVQPY SQEWELVNDD QEPEDIYDDE DDENSENNWR NEYPEEESSD GDEDSRGSAD YNSLSEEERG SSRQRMWSKY PLDVQKEFGY DSPHDLDSD
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADAMS proteinase-related protein; Probable RNA polymerase II nuclear localization protein SLC7A6OS; protein SLC7A6OS; Solute carrier family 7 member 6 opposite strand transcript; solute carrier family 7, member 6 opposite strand
Gene Aliases: SLC7A6OS
UniProt ID: (Human) Q96CW6
Entrez Gene ID: (Human) 84138
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support