Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YINSELQDFQ KVPMQTGILP GCEPCHKKVC AHGTCQPSSQ AGFTCECQEG WMGPLCDQRT NDPCLGNKCV HGTCLPINAF SYSCKCLEGH GGVLCDEEED LFNPCQAIKC KHGKCRLSGL GQPYCECSSG YTGDSCDREI SCRGERIRDY YQKQQGYAAC QTTKKVSRLE C
SLIT2 are ligands for Robo receptors and play a vital role for axonal migration/navigation during neural development. A number of cleavage products through alternate splicing are reported in the literature for SLIT2 proteins. The C-terminal cleavage proteins are more diffusible than the larger N-terminal protein that is more tightly cell associated. SLIT2 protein is expressed in tissues such as fetal lung, kidney, and adult spinal cord.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Slit homolog 2 protein; Slit-2
Gene Aliases: SLIL3; Slit-2; SLIT2
UniProt ID: (Human) O94813
Entrez Gene ID: (Human) 9353
Molecular Function:
immunoglobulin superfamily cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support