Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive test controls include: 293T, PC-3, Mouse lung, Mouse brain, Mouse kidney, Rat brain. The target is usually found in the following locations: Secreted.
Immunogen sequence: GHGGVLCDEE EDLFNPCQAI KCKHGKCRLS GLGQPYCECS SGYTGDSCDR EISCRGERIR DYYQKQQGYA ACQTTKKVSR LECRGGCAGG QCCGPLRSKR R
SLIT2 are ligands for Robo receptors and play a vital role for axonal migration/navigation during neural development. A number of cleavage products through alternate splicing are reported in the literature for SLIT2 proteins. The C-terminal cleavage proteins are more diffusible than the larger N-terminal protein that is more tightly cell associated. SLIT2 protein is expressed in tissues such as fetal lung, kidney, and adult spinal cord.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: downregulated during adipocyte differentiation-1; neurogenic extracellular slit protein; Slit homolog 2 protein; Slit-2
Gene Aliases: Drad-1; E030015M03Rik; E130320P19Rik; mKIAA4141; SLIL3; Slit-2; SLIT2
UniProt ID: (Human) O94813, (Mouse) Q9R1B9
Entrez Gene ID: (Human) 9353, (Rat) 360272, (Mouse) 20563
Molecular Function:
immunoglobulin superfamily cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support