Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SILPAKPFP NSNSMYIDRP PSGKTPQQPP VPPQRPMAAL PPPPAGRNHS PLPPPQTNHE EPSRSRNHKT AKLPAPSIDR STKPPLDRSL APFDREPFTL GKKPPFSDKP SIPAGRSLGE HLPKIQKPPL PPTTERHERS SPLPGKKPPV PKHGWGPDRR ENDEDDVHQR PLPQPALLPM SSNTFPSRST KPSPMNPLPS SHMPGAFSES NSSFPQSASL PPYFSQGPSN RPPIRAEGRN FPLPLPNKPR PPSPAEEENS LNEEWYVSYI TRPEAEAALR KINQDGTFLV RDSSKKTTTN PYVLMVLYKD KVYNIQIRYQ KESQVYLLGT GLRGKEDFLS VSDIIDYFRK MPLLLIDGKN RGSRYQCTLT HAAGYP (159-533 aa encoded by BC016618)
SLP-76 was originally identified as a substrate of the ZAP-70 protein tyrosine kinase following T cell receptor (TCR) ligation in the leukemic T cell line Jurkat. The SLP-76 locus has been localized to human chromosome 5q33 and the gene structure has been partially characterized in mice. The human and murine cDNAs both encode 533 amino acid proteins that are 72% identical and comprised of three modular domains. The NH2-terminus contains an acidic region that includes a PEST domain and several tyrosine residues which are phosphorylated following TCR ligation. SLP-76 also contains a central proline-rich domain and a COOH-terminal SH2 domain. A number of additional proteins have been identified that associate with SLP-76 both constitutively and inducibly following receptor ligation, supporting the notion that SLP-76 functions as an adaptor or scaffold protein. Studies using SLP-76 deficient T cell lines or mice have provided strong evidence that SLP-76 plays a positive role in promoting T cell development and activation as well as mast cell and platelet function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 76 kDa tyrosine phosphoprotein; Lymphocyte cytosolic protein 2; lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of 76kDa); SH2 domain-containing leukocyte protein of 76 kDa; SH2 domain-containing leukocyte protein of 76kD; SLP-76 tyrosine phosphoprotein; SLP76
Gene Aliases: LCP2; SLP-76; SLP76
UniProt ID: (Human) Q13094
Entrez Gene ID: (Human) 3937
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support