Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSATVVDAV NAAPLSGSKE MSLEEPKKMT REDWRKKKEL EEQRKLGNAP AEVDEEGKDI NPHIPQYISS VPWYIDPSKR PTLKHQRPQP EKQKQFSSSG EWYKRGVKEN SVITKYRKGA CENCGAMTHK KKDCFERPRR VGAKFTGTNI APDEHVQPQL MFDYDGKRDR WNGYNPEEHM KIVEEYAKVD LAKRTLKAQK LQEELASGKL VEQANSPKHQ WGEEEPNSQT EKDHNSEDED EDKYADDIDM PGQNFDSKRR ITVRNLRIRE DIAKYLRNLD PNSAYYDPKT RAMRENPYAN AG (1-301 aa encoded by BC010634)
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSlu7; MGC9280; Pre-mRNA-splicing factor SLU7; SLU7 splicing factor homolog; splicing factor; step II splicing factor SLU7; zinc knuckle motif containing
Gene Aliases: 9G8; AU018913; D11Ertd730e; D3Bwg0878e; hSlu7; SLU7
UniProt ID: (Human) O95391, (Mouse) Q8BHJ9, (Rat) Q80ZG5
Entrez Gene ID: (Human) 10569, (Mouse) 193116, (Rat) 303057
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support