Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive test controls include: HeLa, HT-29, Jurkat, NIH/3T3, Rat lung. The target is usually found in the following locations: Cytoplasm, Nucleus.
Immunogen sequence: MSSILPFTPP VVKRLLGWKK SAGGSGGAGG GEQNGQEEKW CEKAVKSLVK KLKKTGRLDE LEKAITTQNC NTKCVTIPST CSEIWGLSTP NTIDQWDTTG
SMAD2 (MADH2, MAD2) regulates multiple cellular processes, such as cell proliferation, apoptosis, and differentiation. SMAD2 interacts with the TGF-beta receptors through its interaction with the SMAD anchor for receptor activation into the nucleus is a central event in TGF beta signaling. Phosphorylation of threonine 8 in the calmodulin-binding region of the MH1 domain by extracellular signal regulated kinase 1 (ERK1) enhances SMAD2 transcriptional activity, which is negatively regulated by calmodulin. In response to the TGF-beta signal, SMAD2 is phosphorylated by the TGF-beta receptors. The phosphorylation induces the dissociation of this protein with SARA and the association with the family member SMAD4. The association with SMAD4 is important for the translocation of this protein into the nucleus, where it binds to target promoters and forms a transcription repressor complex with other cofactors. SMAD2 can also be phosphorylated by activin type 1 receptor kinase, and mediates the signal from the activin. Alternatively spliced transcript variants encoding the SMAD2 protein have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMAD-2; JV18-1; MAD; MAD homolog 2; Mad protein homolog; Mad-related protein 2; MGC22139; MGC34440; mMad2; mother against DPP homolog 2; Mothers against decapentaplegic homolog 2; mothers against DPP homolog 2; pSMAD2; Sma- and Mad-related protein 2; SMAD; SMAD 2; SMAD family member 2; SMAD, mothers against DPP homolog 2; Smad2Deltaexon3
Gene Aliases: 7120426M23Rik; hMAD-2; hSMAD2; JV18; JV18-1; MADH2; MADR2; mMad2; Smad-2; SMAD2
UniProt ID: (Human) Q15796, (Mouse) Q62432, (Rat) O70436
Entrez Gene ID: (Human) 4087, (Mouse) 17126, (Rat) 29357
Molecular Function:
DNA-binding transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support