Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
SMADs are members of the MAD-related family of molecules. MAD-related proteins are a family of intracellular proteins that are essential components in the signaling pathways of the serine/threonine kinase receptors of the transforming growth factor beta superfamily. SMADs can be divided into receptor-regulated SMADs (R-SMADs: SMAD1, 2, 5, 8 and 9), common-mediator SMAD (co-SMAD: SMAD4), and inhibitory SMADs (I-SMADs: SMAD6 and 7). SMAD1, 5, 8 and 9 have high degrees of homology and antibodies are available that recognize sequences common to all of them. SMAD8 and SMAD9 are typically used as alternate names for one another in the literature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781C1895; DKFZp781O1323; hSmad5; MAD homolog 5; Mothers against decapentaplegic homolog 5; mothers against DPP homolog 5; OTTHUMP00000223331; piggytail; Protein somitabun; SMA- and MAD-related protein 5; SMAD; SMAD 5; SMAD family member 5; somitabun; transcription factor
Gene Aliases: fb67b04; mad; madh5; pgy; sb:cb400; SBN; smad5; smd5; wu:fb67b04
UniProt ID: (Zebrafish) Q9W7E7
Entrez Gene ID: (Zebrafish) 30641
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support