Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MFRSKRSGLV RRLWRSRVVP DREEGGSGGG GGGDEDGSLG SRAEPAPRAR EGGGCGRSEV RPVAPRRPRD AVGQRGAQGA GRRRRAGGPP RPMSEPGAGA GSSLLDVAEP GGPGWLPESD CETVTCCLFS ERDAAGAPRD ASDPLAGAAL EPAGGGRSRE ARSRLLLLEQ ELKTVTYSLL KRLKERSLDT LLEAVESRGG VPGGCVLVPR ADLRLGGQPA PPQLLLGRLF RWPDLQHAVE LKPLCGCHSF AAAADGPTVC CNPYHFSRLC GPESPPPPYS RLSPRDEYKP LDLSDSTLSY TETEATNSLI TAPGEFSDAS MSPDATKPSH WCSVAYWEHR TRVGRLYAVY DQAVSIFYDL PQGSGFCLGQ LNLEQRSESV RRTRSKIGFG ILLSKEPDGV WAYNRGEHPI FVNSPTLDAP GGRALVVRKV PPGYSIKVFD FERSGLQHAP EPDAADGPYD PNSVRISFAK GWGPCYSRQF ITSCPCWLEI LLNNPR
SMAD6 (mothers against decapentaplegic homolog 6) is an antagonist of signaling by TGF-beta (transforming growth factor) type 1 receptor superfamily members; has been shown to inhibit selectively BMP (bone morphogenetic proteins) signaling by competing with the co-SMAD SMAD4 for receptor-activated SMAD1. SMAD6 is an inhibitory SMAD (I-SMAD) or antagonistic SMAD.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MAD homolog 6; Mothers against decapentaplegic homolog 6; mothers against decapentaplegic, drosophila, homolog of, 6; SMAD; SMAD 6; SMAD family member 6; SMAD SMAD; SMAD, mothers against DPP homolog 6
Gene Aliases: AOVD2; HsT17432; MADH6; MADH7; SMAD6
UniProt ID: (Human) O43541
Entrez Gene ID: (Human) 4091
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support