Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MYIKQVIIQ GFRSYRDQTI VDPFSSKHNV IVGRNGSGKS NLFYAIQFVL SDEFSHLRPE QRLALLHEGT GPRVISAFVE IIFDNSDNRL PIDKEEVSLR RVIGAKKDQY FLDKKMVTKN DVMNLLESAG FSRSNPYYIV KQGKINQMAT APDSQRLKLL REVAGTRVYD ERKEESISLM KETEGKREKI NELLKYIEER LHTLEEEKEE LAQYQKWDKM RRALEYTIYN QELNETRAKL DELSAKRETS GEKSRQLRDA QQDARDKMED IERQVRELKT KISAMKEEKE QLSAERQEQI KQRTKLELKA KDLQDELAGN SEQRKRLLKE RQKLLEKIEE KQKELAETEP (1-349 aa encoded by BC047324)
This gene belongs to the SMC3 subfamily of SMC proteins. The encoded protein occurs in certain cell types as either an intracellular, nuclear protein or a secreted protein. The nuclear form, known as structural maintenance of chromosomes 3, is a component of the multimeric cohesin complex that holds together sister chromatids during mitosis, enabling proper chromosome segregation. Post-translational modification of the encoded protein by the addition of chondroitin sulfate chains gives rise to the secreted proteoglycan bamacan, an abundant basement membrane protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bamacan; basement membrane chondroitin sulfate proteoglycan; Basement membrane-associated chondroitin proteoglycan; Chondroitin sulfate proteoglycan 6; chondroitin sulfate proteoglycan 6 (bamacan); Chromosome segregation protein SmcD; Chromosome-associated polypeptide; hCAP; Mad member-interacting protein 1; SMC protein 3; structural maintenace of chromosomes 3; Structural maintenance of chromosomes protein 3
Gene Aliases: BAM; Bamacan; BMH; CDLS3; CSPG6; HCAP; Mmip1; SMC-3; SMC3; SMC3L1; SmcD
UniProt ID: (Human) Q9UQE7, (Mouse) Q9CW03
Entrez Gene ID: (Human) 9126, (Mouse) 13006, (Rat) 29486
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support