Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAQKGQLSD DEKFLFVDKN FINSPVAQAD WAAKRLVWVP SEKQGFEAAS IKEEKGDEVV VELVENGKKV TVGKDDIQKM NPPKFSKVED MAELTCLNEA SVLHNLRERY FSGLIYTYSG LFCVVVNPYK HLPIYSEKIV DMYKGKKRHE MPPHIYAIAD TAYRSMLQDR EDQSILCTGE SGAGKTENTK KVIQYLAVVA SSHKGKKDTS ITGELEKQLL QANPILEAFG NAKTVKNDNS SRFGKFIRIN FDVTGYIVGA NIETYLLEKS RAIRQARDER TFHIFYYMIA GAKEKMRSDL LLEGFNNYTF LSNGFVPIPA AQDDEMFQET VEAMAIMGFS EEEQLSILKV VS (1-351 aa encoded by BC101677)
SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: myh11 {ECO:0000312|RGD:3136}; Myosin heavy chain 11; myosin heavy chain 11, smooth muscle; myosin heavy chain 21; Myosin heavy chain, smooth muscle isoform; myosin, heavy chain 11, smooth muscle; myosin, heavy polypeptide 11, smooth muscle; Myosin-11; SMMHC; smooth muscle myosin heavy chain
Gene Aliases: AAT4; AV071570; FAA4; KIAA0866; MYH11; SM1; SM2; SMHC; SMMHC
UniProt ID: (Human) P35749
Entrez Gene ID: (Human) 4629, (Rat) 24582, (Mouse) 17880
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support