Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAMSSGGSG GGVPEQEDSV LFRRGTGQSD DSDIWDDTAL IKAYDKAVAS FKHALKNGDI CETSGKPKTT PKRKPAKKNK SQKKNTAASL QQWKVGDKCS AIWSEDGCIY PATIASIDFK RETCVVVYTG YGNREEQNLS DLLSPICEVA NNIEQNAQEN ENESQVSTDE SENSRSPGNK SDNIKPKSAP WNSFLPPPPP MPGPRLGPGK PGLKFNGPPP PPPPPPPHLL SCWLPPFPSG PPIIPPPPPI CPDSLDDADA LGSMLISWYM SGYHTGYYME MLA (1-282 aa encoded by BC000908)
The SMN1 gene is part of a 500 kb inverted duplication on chromosome 5q13. This duplicated region contains at least four genes and repetitive elements which make it prone to rearrangements and deletions. The repetitiveness and complexity of the sequence have also caused difficulty in determining the organization of this genomic region. The telomeric and centromeric copies of this gene are nearly identical and encode the same protein - survival motor neuron protein. The SMN complex plays a catalyst role in the assemble of small nuclear ribonucleoproteins, the building blocks of the spliceosome. Mutations in the SMN1 gene are known to cause spinal muscular atrophy 1/2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Component of gems 1; Gemin-1; survival motor neuron 1 protein; Survival motor neuron protein; survival of motor neuron protein; tudor domain containing 16A
Gene Aliases: AI849087; BCD541; GEMIN1; SMA; SMA1; SMA2; SMA3; SMA4; SMA@; SMN; SMN1; SMN2; SMNC; SMNT; T-BCD541; TDRD16A
UniProt ID: (Human) Q16637, (Mouse) P97801
Entrez Gene ID: (Human) 6606, (Mouse) 20595
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support