Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 4
Positive Control: HeLa cell lysate.
Tissue Specificity: Widely expressed in many tissues with a strongest signal in ovary. No expression in spleen.
Subcellular Location: Secreted, extracellular space, extracellular matrix, basement membrane.
Sequence Similarities: No Cross with Mouse/Rat.
Predicted band size: 223 kDa.
Plays essential roles in both eye and limb development. Probable regulator of osteoblast differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Secreted modular calcium-binding protein 1; SMOC-1; SPARC-related modular calcium-binding protein 1
Gene Aliases: OAS; SMOC1
UniProt ID: (Human) Q9H4F8
Entrez Gene ID: (Human) 64093
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support