Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
SMUG1 is a glycosylase that removes uracil from single- and double-stranded DNA in nuclear chromatin, thus contributing to base excision repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC104370; Single-strand selective monofunctional uracil DNA glycosylase; Single-strand selective monofunctional uracil-DNA glycosylase
Gene Aliases: 1200013B09Rik; A930006H09Rik; C85220; FDG; HMUDG; SMUG1; UNG3
UniProt ID: (Human) Q53HV7, (Mouse) Q6P5C5, (Rat) Q811Q1
Entrez Gene ID: (Human) 23583, (Mouse) 71726, (Rat) 315344
Molecular Function:
DNA metabolism protein
DNA glycosylase
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support