Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AGHCRIEVSR EEIFEESYRQ VMKMRPKDLW KRLMIKFRGE EGLDYGGVAR EWLYLLSHEM LNPYYGLFQY SRDDIYTLQI NPDSAVNPEH LSYFHFVGRI MGMAVFHGHY IDGGFTLPFY KQLLGKSITL DDMELVDPDL HNSLVWILEN DITGVLDHTF CVEHNAYGEI IQHELKPNGK SIPVNEENKK EYVRLYVNWR FLRGIEAQFL ALQKGFNEVI PQHLLKTFDE KELELIICGL GKIDVNDWKV NTRLKHCTPD SNIVKWFWKA VEFFDEERRA RLLQFVTGSS RVPLQGFKAL QGAAGPRLFT IHQIDACTNN LPKAHTCFNR IDIPPYESYE KLYEKLLTAI EETCGFAVE
SMURF2 is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. This protein interacts with SMAD1, SMAD2 and SMAD7 in order to trigger their ubiquitination and proteasome-dependent degradation. It enhances the inhibitory activity of SMAD7 and reduces the transcriptional activity of SMAD2. Coexpression of SMURF2 with SMAD1 results in considerable decrease in steady-state level of SMAD1 protein and a smaller decrease of SMAD2 level.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin ligase SMURF2; E3 ubiquitin-protein ligase SMURF2; HECT-type E3 ubiquitin transferase SMURF2; hSMURF2; SMAD ubiquitination regulatory factor 2; SMAD-specific E3 ubiquitin-protein ligase 2
Gene Aliases: 2810411E22Rik; AI558114; AI649275; RGD1310067; SMURF2
UniProt ID: (Human) Q9HAU4, (Mouse) A2A5Z6
Entrez Gene ID: (Human) 64750, (Rat) 303614, (Mouse) 66313
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support