Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Acts as a transcriptional repressor. Essential for cardiomyocyte differentiation and cardiac morphogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD8 beta opposite; CD8b-opposite; CD8beta opposite strand; Histone-lysine N-methyltransferase SMYD1; m-BOP; SET and MYND domain-containing protein 1; Zinc finger protein BOP; zinc finger, MYND domain containing 18
Gene Aliases: 4632404M21Rik; BOP; C78565; KMT3D; SMYD1; ZMYND18; ZMYND22
UniProt ID: (Human) Q8NB12, (Mouse) P97443
Entrez Gene ID: (Human) 150572, (Mouse) 12180
Molecular Function:
histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support