Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SMYD3 (histone-lysine N-methyltransferase) methylates Lys-4 of histone H3 and Lys-5 of histone H4. It induces di- and tri-methylation. SMYD3 plays an important role in transcriptional activation as a member of an RNA polymerase complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA74P14.1 (novel protein); Histone-lysine N-methyltransferase SMYD3; SET and MYND domain-containing protein 3; Zinc finger MYND domain-containing protein 1; zinc finger protein, subfamily 3A (MYND domain containing), 1; zinc finger, MYND domain containing 1
Gene Aliases: 2410008A19Rik; bA74P14.1; KMT3E; SMYD3; ZMYND1; ZNFN3A1
UniProt ID: (Human) Q9H7B4, (Mouse) Q9CWR2
Entrez Gene ID: (Human) 64754, (Mouse) 69726, (Rat) 498295
Molecular Function: histone modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support