Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Positive Samples: HepG2, A-549, Mouse brain, Rat brain
Immunogen sequence: SAKDYFFKAA LCHFCIDMLN AKLAVQKYEE LFPAFSDSRE CKLMKKLLEA HEEQNVDSYT ESVKEYDSIS RLDQWLTTML LRIKKTIQGD EEDLR
SNAPs (soluble NSF attachment proteins), acting in concert with SNAREs (SNAP receptors) and the N-ethylmaleimide-sensitive fusion protein (NSF) are required for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-to-endosome fusion and transcytotic vesicles-to-plasma membrane transport. Vesicle-to-target membrane docking (initial contact) occurs when the vesicle SNARE binds to its cognate target membrane SNARE. a-SNAP (or b-SNAP in brain) then binds to this docking complex and mediates the binding of NSF and thus the formation of a 20S fusion particle. It is thought that, once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha snap; alpha soluble NSF attachment protein; alpha-SNAP; Alpha-soluble NSF attachment protein; hydrocephaly with hop gait; N-ethylmaleimide sensitive fusion protein attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein, alpha; SNAP-alpha
Gene Aliases: 1500039N14Rik; a-SNAP; alpha-SNAP; AW209189; hyh; NAPA; RA81; Snap; SNAPA; SNARE
UniProt ID: (Human) P54920, (Rat) P54921, (Mouse) Q9DB05
Entrez Gene ID: (Human) 8775, (Rat) 140673, (Mouse) 108124
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support