Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MSAYPKSYNP FDDDGEDEGA RPAPWRDARD LPDGPDAPAD RQQYLRQEVL RRAEATAAST SRSLALMYES EKVGVASSEE LARQRGVLER TEKMVDKMDQ DLKISQKHIN SIKSVFGGLV NYFKSKPVET PPEQNGTLTS QPNNRLKEAI STSKEQEAKY QASHPNLRKL DDTDPVPRGA GSAMSTDAYP KNPHLRAYHQ KIDSNLDELS MGLGRLKDIA LGMQTEIEEQ DDILDRLTTK VDKLDVNIKS TERKVRQL
SNAP29 encodes a product that is a member of the SNAP25 gene family, encodes a protein involved in multiple membrane trafficking steps. Two other members of this gene family, SNAP23 and SNAP25, encode proteins that bind a syntaxin protein and mediate synaptic vesicle membrane docking and fusion to the plasma membrane. The protein encoded by this gene binds tightly to multiple syntaxins and is localized to intracellular membrane structures rather than to the plasma membrane. While the protein is mostly membrane-bound, a significant fraction of it is found free in the cytoplasm. Use of multiple polyadenylation sites has been noted for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ21051; SNAP-29; Soluble 29 kDa NSF attachment protein; Synaptosomal-associated protein 29; synaptosomal-associated protein, 29kD; synaptosomal-associated protein, 29kDa; synaptosome associated protein 29kDa; Vesicle-membrane fusion protein SNAP-29
Gene Aliases: CEDNIK; SNAP-29; SNAP29
UniProt ID: (Human) O95721
Entrez Gene ID: (Human) 9342
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support