Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MDVFMKGLSM AKEGVVAAAE KTKQGVTEAA EKTKEGVLYV GSKTREGVVQ GVASVAEKTK EQASHLGGAV FSGAGNIAAA TGLVKREEFP TDLKPEEVAQ EAAEEPLIEP LMEPEGESYE DPPQEEYQEY EPEA
Synuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's PD) and breast cancer. In AD, a peptide derived from alpha synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of alpha synuclein has been linked to many familial cases of PD. In breast cancer increased expression of gamma synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-synuclein
Gene Aliases: SNCB
UniProt ID: (Human) Q16143
Entrez Gene ID: (Human) 6620
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support