Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: ERGTVLAEDQ LAQMSKQLDM FKTNLEEFAS KHKQEIRKNP EFRVQFQDMC ATIGVDPLAS GKGFWSEMLG VGDFYYELGV QIIEVCLALK HRNGGLITLE
ELL encodes an RNA polymerase II transcription factor that undergoes frequent translocation in acute myeloid leukemia (AML). In addition to its elongation activity, ELL contains a novel type of RNA polymerase II interaction domain that is capable of repressing polymerase activity in promoter-specific transcription. EAP30 is a subunit of the ELL complex. EAP30 can interact with ELL and derepress ELL's inhibitory activity in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EAP30 subunit of ELL complex; ELL-associated protein of 30 kDa; ESCRT-II complex subunit VPS22; hVps22; SNF8, ESCRT-II complex subunit, homolog; Vacuolar-sorting protein SNF8
Gene Aliases: Dot3; EAP30; SNF8; VPS22
UniProt ID: (Human) Q96H20
Entrez Gene ID: (Human) 11267
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support