Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EGHIVTCET NTGEVYRGKL IEAEDNMNCQ MSNITVTYRD GRVAQLEQVY IRGSKIRFLI LPDMLKNAPM LKSMKNKNQG SGAGRGKAAI LKAQVAARGR GRGMGRG (14-119 aa encoded by BC000457)
This gene encodes a core component of the spliceosome, which is a nuclear ribonucleoprotein complex that functions in pre-mRNA splicing. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sm-D3; small nuclear ribonucleoprotein D3 polypeptide 18kDa; Small nuclear ribonucleoprotein Sm D3; snRNP core protein D3
Gene Aliases: Sm-D3; SMD3; SNRPD3
UniProt ID: (Human) P62318
Entrez Gene ID: (Human) 6634
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support