Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSLPLNPKP FLNGLTGKPV MVKLKWGMEY KGYLVSVDGY MNMQLANTEE YIDGALSGHL GEVLIRCNNV LYIRGVEEEE EDGEMRE (1-86 aa encoded by B C002505)
SNRPF belongs to the snRNP Sm proteins family and is associated with snRNP U1, U2, U4/U6 and U5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sm protein F; Sm-F; Small nuclear ribonucleoprotein F; snRNP-F
Gene Aliases: 2010013O18Rik; PBSCF; Sm-F; SMF; snRNP-F; SNRPF
UniProt ID: (Human) P62306, (Mouse) P62307
Entrez Gene ID: (Human) 6636, (Mouse) 69878, (Rat) 680737
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support