Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: VRVVKQEAGG LGISIKGGRE NRMPILISKI FPGLAADQSR ALRLGDAILS VNGTDLRQAT HDQAVQALKR AGKEVLLEVK FIREVTPYIK KPSLV
Syntrophins are structurally related proteins located at the neuromuscular synapse and play an important role in dystrophin associated protein (DAP) complex formation. To date there have been three distinct syntrophin isoforms discovered: alpha 1, beta 1, and beta 2. These proteins each contain a PDZ domain, a syntrophin-unique domain (SU), and three pleckstrin homology domains (PH1a,1b and 2). The PDZ domain has been shown to be responsible for binding syntrophin to nNOS and the DAP complex. This association results in localization of the nicotinic acetylcholine receptor to the cytoskeleton via the (S/T)XV C terminus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 59 kDa dystrophin-associated protein A1 basic component 2; Beta-2-syntrophin; dystrophin-associated protein A1, 59kD, basic component 2; SNT3; SNTL; SYN; syntrophin, beta 2 (dystrophin-associated protein A1, 59kDa, basic component 2); Syntrophin-3; Syntrophin-like
Gene Aliases: D16S2531E; EST25263; SNT2B2; SNT3; SNTB2; SNTL
UniProt ID: (Human) Q13425
Entrez Gene ID: (Human) 6645
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support