Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: MHFSIPETES RSGDSGGSAY VAYNIHVNGV LHCRVRYSQL LGLHEQLRKE YGANVLPAFP PKKLFSLTPA EVEQRREQLE KYMQAVRQDP LLGSSETFNS FLRRAQQETQ QVPTEEVSLE VLLSNGQKVL VNVLTSDQTE DVLEAVAAKL DLPDDLIGYF SLFLVREKED GAFSFVRKLQ EFELPYVSVT SLRSQEYKIV LRKSYWDSAY DDDVMENRVG LNLLYAQTVS DIERGWILVT KEQHRQLKSL QEKVSKKEFL RLAQTLRHYG YLRFDACVAD FPEKDCPVVV SAGNSELSLQ LRLPGRQLRE GSFRVTRMRC WRVTSSVPLP SGSTSSPGRG RGEVRLELAF EYLMSKDRLQ WVTITSPQAI MMSICLQSMV DELMVKKSGG SIRKMLRRRV GGTLRRSDSQ QAVKSPPLLE SPDATRESMV KLSSKLSAVS LRGIGSPSTD ASASDVHGNF AFEGIGDEDL
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a B41 domain. This protein interacts with the cytoplasmic domain of P-selectin, and may function in the intracellular trafficking of P-selectin. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sorting nexin-17
Gene Aliases: KIAA0064; SNX17
UniProt ID: (Human) Q15036
Entrez Gene ID: (Human) 9784
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support