Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
May be involved in several stages of intracellular trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTB-36H16.2; MGC5204; Sorting nexin-2; Transformation-related gene 9 protein; TRG-9
Gene Aliases: 0610030A03Rik; SNX2; TRG-9; TRG9
UniProt ID: (Human) Q53XK8, (Mouse) Q9CWK8
Entrez Gene ID: (Human) 6643, (Mouse) 67804
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support