Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VERRRIGLE NFLLRIASHP ILCRDKIFYL FLTQEGNWKE TVNETGFQLK ADSRLKALNA TFRVKNPDKR FTDLKHYSDE LQSVISHLLR VRARVADRLY GVYKVHGNYG RVFSEWSAIE KEMGDGLQSA GHHMDVYASS IDDILEDEEH YADQLKEYLF YAEALRAVCR KHELMQYDLE MAAQDLASKK QQCEELVTGT VRTFSLKGMT TKLFGQETPE QREARIKVLE EQINEGEQQL KSKNLEGREF VKNAWADIER FKEQKNRDLK EALISYAVMQ ISMCKKGIQV WTNAKECFSK M (151-450 aa encoded by BC018762)
SNX2 and SNX4 share a high degree of amino acid similarity with SNX1, as they all contain a characteristic phox homology (PX) domain. These proteins are all partially associated with cellular membranes, and they, likewise, associate with EGF, PDGF and insulin receptor tyrosine kinases. These nexins are widely expressed and yet have various tissue distribution patterns. Additionally, the sorting nexins can associate with each other and with a variety of other cellular proteins, suggesting that they exist as part of multisubunit complexes. The related protein, SNX3, comprises a distinct subgroup of nexins that share less sequence similarity outside of the PX domain and have dramatically different binding affinities for the tyrosine kinase receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Sorting nexin-4
Gene Aliases: 1810036H14Rik; AI037066; ATG24B; SNX4
UniProt ID: (Human) O95219, (Mouse) Q91YJ2
Entrez Gene ID: (Human) 8723, (Mouse) 69150, (Rat) 360725
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support