Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAVPELLQ QQEEDRSKLR SVSVDLNVDP SLQIDIPDAL SERDKVKFTV HTKTTLPTFQ SPEFSVTRQH EDFVWLHDTL IETTDYAGLI IPPAPTKPDF DGPREKMQKL GEGEGSMTKE EFAKMKQELE AEYLAVFKKT VSSHEVFLQR LSSHPVLSKD RNFHVFLEYD QDLSVRRKNT KEMFGGFFKS VVKSADEVLF TGVKEVDDFF EQEKNFLINY YNRIKDSCVK ADKMTRSHKN VADDYIHTAA CLHSLALEEP TVIKKYLLKV AELFEKLRKV EGRVSSDEDL KLTELLRYYM LNIEAAKDLL YRRTKALIDY ENSNKALDKA RLKSKDVKLA EAHQQECCQK FEQLSESAKE ELINFKRKRV AAFRKNLIEM SELEIKHARN NVSLLQSCID LFKNN (1-404 aa encoded by BC093623)
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein binds to fanconi anemia complementation group A protein, but its function is unknown. This gene results in two transcript variants encoding the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10931; RP11-504H3.2; Sorting nexin-5
Gene Aliases: 0910001N05Rik; 1810032P22Rik; AU019504; D2Ertd52e; SNX5
UniProt ID: (Human) Q9Y5X3, (Mouse) Q9D8U8
Entrez Gene ID: (Human) 27131, (Rat) 296199, (Mouse) 69178
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support