Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAENNENISK NVDVRPKTSR SRSADRKDGY VWSGKKLSWS KKSESYSDAE TVNGIEKTEV SLRNQERKHS CSSIELDLDH SCGHRFLGRS LKQKLQDAVG QCFPIKNCSS RHSSGLPSKR KIHISELMLD KCPFPPRSDL AFRWHFIKRH TAPINSKSDE WVSTDLSQTE LRDGQLKRRN MEENINCFSH TNVQPCVITT DNALCREGPM TGSVMNLVSN NSIEDSDMDS DDEILTLCTS SRKRNKPKWD LDDEILQLET PPKYHTQIDY VHCLVPDLLQ INNNPCYWGV MDKYAAEALL EGKPEGTFLL RDSAQEDYLF SVSFRRYSRS LHARIEQWNH NFSFDAHDPC VFHSPDITGL LEHYKDPSAC MFFEPLLSTP LIRTFPFSLQ HICRTVICNC TTYDGIDALP IPSSMKLYLK EYHYKSKVRV LRIDAPEQQC
The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SH2 domain containing SOCS box protein; SOCS-4; SOCS-7; Suppressor of cytokine signaling 4; Suppressor of cytokine signaling 7
Gene Aliases: SOCS4; SOCS7
UniProt ID: (Human) Q8WXH5
Entrez Gene ID: (Human) 122809
Molecular Function: kinase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support