Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Accumulating evidence demonstrates that cytokine receptor signaling is negatively regulated by a family of Src homology 2 domain-containing adaptor molecules Termed SOCS (Suppressor of Cytokine Signaling). To date, there are eight members of SOCS family have been recognized, they are SOCS-1, 2, 3, 4, 5, 6, 7 and CIS. Structurally the SOCS proteins are composed of an N-terminal region of variable length and amino acid composition, a central SH2 domain, and a previously unrecognized C-terminal motif that we have called the SOCS box. The SOCS proteins appear to form part of a classical negative feed back loop that regulates cytokine signal transduction via a STAT-induced transcriptional mechanism. Transcription of each of the SOCS genes occurs rapidly in vitro and in vivo in response to cytokines, and once produced, the various members of the SOCS family appear to inhibit signaling in different ways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CIS-6; cytokine inducible SH2-containing protein 5; Cytokine-inducible SH2 protein 6; Cytokine-inducible SH2-containing protein 5; SOCS-5; Suppressor of cytokine signaling 5
Gene Aliases: 1810018L08Rik; CIS6; CISH5; CISH6; KIAA0671; mKIAA0671; RGD1564914; SOCS-5; SOCS5
UniProt ID: (Human) O75159, (Mouse) O54928
Entrez Gene ID: (Human) 9655, (Rat) 500616, (Mouse) 56468
Molecular Function: kinase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support