Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: LDPKALPPGL ALERTWGPAA GLEAQLAALG LGQPAGPGVK TVGGGCCPCP
Sequence homology: Cow: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
SOCS7 regulates signaling cascades probably through protein ubiquitination and/or sequestration. SOCS7 functions in insulin signaling and glucose homeostasis through IRS1 ubiquitination and subsequent proteasomal degradation. SOCS7 inhibits also prolactin, growth hormone and leptin signaling by preventing STAT3 and STAT5 activation, sequestering them in the cytoplasm and reducing their binding to DNA. SOCS7 may be a substrate recognition component of a SCF-like E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NAP-4; Nck, Ash and phospholipase C binding protein; Nck, Ash and phospholipase C gamma-binding protein; NCK-associated protein 4; SOCS-7; Suppressor of cytokine signaling 7
Gene Aliases: NAP4; NCKAP4; SOCS6; SOCS7
UniProt ID: (Human) O14512
Entrez Gene ID: (Human) 30837
Molecular Function:
kinase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support