Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MVQQAESLEA ESNLPREALD TEEGEFMACS PVALDESDPD WCKTASGHIK RPMNAFMVWS KIERRKIMEQ SPDMHNAEIS KRLGKRWKML KDSEKIPFIR
This intronless gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may function in the developing nervous system and play a role in tumorigenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SRY; SRY (sex-determining region Y)-box 11; SRY-box 11; SRY-box containing gene 11; SRY-related HMG-box gene 11; Transcription factor SOX-11
Gene Aliases: 1110038H03Rik; 6230403H02Rik; AI836553; end1; MRD27; Sox-11; SOX11
UniProt ID: (Human) P35716, (Mouse) Q7M6Y2, (Rat) P0C1G9
Entrez Gene ID: (Human) 6664, (Mouse) 20666, (Rat) 84046
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support