Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. It has also been determined to be a type-1 diabetes autoantigen, also known as islet cell antibody 12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: islet cell antibody 12; Islet cell antigen 12; MGC117216; RP11-74C13.1; SRY (Sex determining region Y)-box 13; SRY box 13; SRY-related HMG-box gene 13; Transcription factor SOX-13; Type 1 diabetes autoantigen ICA12
Gene Aliases: ICA12; Sox-13; SOX13
UniProt ID: (Human) Q9UHW7
Entrez Gene ID: (Human) 9580
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support