Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: KSEPSSPPPA IASHSQRACL GDLRDMISMY LPPGGDAADA ASPLPGGRLH GVHQHYQGAG TAVNGTVPLT H*
This gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene have been associated with X-linked mental retardation with growth hormone deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SRY (sex determining region Y)-box 3; SRY box 3; Transcription factor SOX-3
Gene Aliases: GHDX; MRGH; PHP; PHPX; SOX3; SOXB
UniProt ID: (Human) P41225
Entrez Gene ID: (Human) 6658
Molecular Function:
HMG box transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support