Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Transcriptional activator. Binds specifically to the DNA sequence 5'-AACAAT-3'. Plays a key role in several developmental processes, including neurogenesis and skeleton formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box containing gene 6; Transcription factor SOX-6
Gene Aliases: AI987981; HSSOX6; Sox-6; SOX-LZ; SOX6; SOXD
UniProt ID: (Human) P35712, (Rat) A0A0G2JTZ2, (Mouse) P40645
Entrez Gene ID: (Human) 55553, (Rat) 293165, (Mouse) 20679
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support