Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NLQLQAVPN SGPIIIRTPT VGPNGQVSWQ TLQLQNLQVQ NPQAQTITLA PMQGVSLGQT SSSNTTLTPI ASAASIPAGT VTVNAAQLSS MPGLQTINLS ALGTSGIQVH PIQGLPLAIA NAPGDHGAQL GLHGAGGDGI HDDTAGGEEG ENSPDAQPQA GRRTRREACT CPYCKDSEGR GSGDPGKKKQ HICHIQGCGK VYGKTSHLRA HLRWHTGERP FMCTWSYCGK RFTRSDELQR HKRTHTGEKK FACPECPKRF MRSDHLSKHI KTHQNKKGGP GVALSVGTLP LDSGAGSEGS GTATPSALIT TNMVAMEAIC PEGIARLANS GINVMQVADL QSINISGNGF (437-785 aa encoded by BC062539)
SP1 is a transcription factor that can activate or repress transcription in response to physiological and pathological stimuli. Binds with high affinity to GC-rich motifs and regulates the expression of a large number of genes involved in a variety of processes such as cell growth, apoptosis, differentiation and immune responses. Highly regulated by post-translational modifications (phosphorylations, sumoylation, proteolytic cleavage, glycosylation and acetylation). Binds also the PDGFR-alpha G-box promoter. May have a role in modulating the cellular response to DNA damage. Implicated in chromatin remodeling. Plays a role in the recruitment of SMARCA4/BRG1 on the c-FOS promoter. Plays an essential role in the regulation of FE65 gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:11205; Specificity protein 1; trans-acting transcription factor 1; Transcription factor Sp1
Gene Aliases: 1110003E12Rik; AA450830; AI845540; SP1; Sp1-1; TSFP1
UniProt ID: (Human) P08047, (Mouse) O89090, (Rat) Q01714
Entrez Gene ID: (Human) 6667, (Mouse) 20683, (Rat) 24790
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support