Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: MASSLLEEEV HYGSSPLAML TAACSKFGGS SPLRDSTTLG KAGTKKPYSV
Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 86%
SP7 is a C2H2-type zinc finger transcription factor of the SP gene family and a putative master regulator of bone cell differentiation (Gao et al., 2004 [PubMed 15474293]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC126598; Transcription factor Sp7; Zinc finger protein osterix
Gene Aliases: OI11; OI12; osterix; OSX; SP7
UniProt ID: (Human) Q8TDD2
Entrez Gene ID: (Human) 121340
Molecular Function:
C2H2 zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support