Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: AELDKLHKEV STVRAANSER VAKLVFQRLN EDFVRKPDYA LSSVGASIDL QKTSHDYADR NTAYFWNRFS FWNYARPPTV ILEPHVFPGN CWAFEGDQGQ VVIQLPGRVQ
The mammalian sperm flagellum contains two cytoskeletal structures associated with the axoneme: the outer dense fibers surrounding the axoneme in the midpiece and principal piece and the fibrous sheath surrounding the outer dense fibers in the principal piece of the tail. Defects in these structures are associated with abnormal tail morphology, reduced sperm motility, and infertility. In the rat, the protein encoded by this gene associates with an outer dense fiber protein via a leucine zipper motif and localizes to the microtubules of the manchette and axoneme during sperm tail development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acrosomal protein ACR55; cancer/testis antigen 127; Outer dense fiber-associated protein SPAG4; Sad1 and UNC84 domain containing 4; sperm tail protein; Sperm-associated antigen 4 protein; SUN domain-containing protein 4
Gene Aliases: CT127; SPAG4; SUN4
UniProt ID: (Human) Q9NPE6
Entrez Gene ID: (Human) 6676
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support