Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SPATA22 is highly expressed in adult testis. The specific function of SPATA22 is not yet known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Spermatogenesis-associated protein 22; testicular tissue protein Li 186; Testis development protein NYD-SP20
Gene Aliases: Gm882; NYD-SP20; NYDSP20; Repro42; SPATA22
UniProt ID: (Human) Q8NHS9, (Mouse) Q5SV06
Entrez Gene ID: (Human) 84690, (Mouse) 380709
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support