Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAAAVQGG RSGGSGGCSG AGGASNCGTG SGRSGLLDKW KIDDKPVKID KWDGSAVKNS LDDSVKKVLL EKYKYVENFG LIDGRLTICT ISCFFAIVAL IWDYMHPFPE SKPVLALCVI SYFVMMGILT IYTSYKEKSI FLVAHRKDPT GMDPDDIWQL SSSLKRFDDK YTLKLTFISG RTKQQREAEF TKSIAKFFDH SGTLVMDAYE PEISRLHDSL AIERKIK (1-226 aa encoded by BC064957)
Component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC117366; Microsomal signal peptidase 25 kDa subunit; signal peptidase 25kDa subunit; Signal peptidase complex subunit 2; signal peptidase complex subunit 2 homolog; SPase 25 kDa subunit
Gene Aliases: 5730406I15Rik; AA408713; KIAA0102; mKIAA0102; SPC25; SPCS2
UniProt ID: (Human) Q15005, (Mouse) Q9CYN2
Entrez Gene ID: (Human) 9789, (Mouse) 66624
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support