Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TLVGMKLLD KISSVPHGEL SCTTELLILA HHCFTLTCHM EGIIRVLQAA HMLTDNHLAP SEEYGLVVRL LTGIGRYNEM TYIFDLLHKK HYFEVLMRKK LDPSGTLKTA LLDYIKRCRP GDSEKHNMIA LCFSMCREIG ENHEAAARIQ LKLIESQPWE DSLKDGHQLK QLLLKALTLM LDAAESYAKD SCVRQAQHCQ RLTKLITLQI HFLNTGQNTM LINLGRHKLM DCILALPRFY QASIVAEAYD FVPDWAEILY QQVILKGDFN YLEEFKQQRL LKSSIFEEIS KKYKQHQPTD MVMENLKKLL TYCEDVYLYY KLAYEHKFYE IVNVLLKDPQ TGCCLKDMLA G (133-482 aa encoded by BC024161)
Hereditary spastic paraplegias (HSPs) are genetically and phenotypically heterogeneous disorders. Spastic paraplegia with thinning of the corpus callosum (ARHSP-TCC) is a relatively frequent form of complicated hereditary spastic paraplegia (cHSP) in which mental retardation and muscle stiffness at onset are followed by slowly progressive paraparesis and cognitive deterioration. Mutations of the SPG11 gene encoding the spatacsin protein have been identified as a major cause of HSP-TCC. Spatacsin is a potential transmembrane protein that is phosphorylated upon DNA damage. It is expressed in all structures of the brain, with a high expression in the cerebellum. SPG11 mutations may occur more frequently in familial than sporadic forms of cHSP without TCC. Kjellin syndrome is found to be associated with mutations in not only the SPG15 gene but also SPG11 gene. Recent studies show Parkinsonism may initiate SPG11-linked HSP TCC and that SPG11 may cause juvenile Parkinsonism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Colorectal carcinoma-associated protein; Spastic paraplegia 11 protein; Spastic paraplegia 11 protein homolog; Spatacsin
Gene Aliases: 6030465E24Rik; A330015I11; ALS5; C530005A01Rik; CMT2X; KIAA1840; RGD1562529; SPG11
UniProt ID: (Human) Q96JI7, (Mouse) Q3UHA3
Entrez Gene ID: (Human) 80208, (Mouse) 214585, (Rat) 311372
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support